Enalapril-Maleat
Lieferant: ALFA AESAR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
J63932.06
J63932.06EA 203 EUR InStock
J63932.06
Enalapril-Maleat
Enalapril maleate
CAS No.: 76095-16-4 Molecular Formula: C20H28N2O5.C4H4O4 Formula Weight: 492.52 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to off-white Melting Point: 143-145° MDL No.: MFCD00133304
Applications: Long-acting ACE inhibitor and antihypertensive
Formel: C₂₀H₂₈N₂O₅
MW: 492.53 g/mol
Melting Pt: 143…145 °C
MDL Number: MFCD00133304
CAS-Nummer: 76095-16-4
EINECS: 278-375-7
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge