5-Iodocytidine 99%
Lieferant: ALFA AESAR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
J64698.03
J64698.03EA 54.3 EUR InStock
J64698.03
5-Iodocytidine 99%
5-Iodocytidine
Purity: 99% CAS No.: 1147-23-5 Molecular Formula: C9H12IN3O5 Formula Weight: 369.11 Storage Temperature: 2°C to 8°C Physical Form: Crystalline or powder Sensitivity: Light Sensitive Appearance: White to off-white MDL No.: MFCD00056070
Formel: C₉H₁₂IN₃O₅
MDL Number: MFCD00056070
CAS-Nummer: 1147-23-5
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge