Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Guanidiniumthiocyanat für die Molekularbiologie

Lieferant: ALFA AESAR

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

J65104.18 J65104.30 J65104.36
J65104.18EA 80.8 EUR InStock
J65104.18 J65104.30 J65104.36
Guanidiniumthiocyanat für die Molekularbiologie
Guanidiniumthiocyanat
Guanidine thiocyanate is used for RNA isolation and can solubilise proteins. When the compound is used to solubilize proteins it can be removed by dialysis against urea without difficulty. The hybridization of guanidine thiocyanate-solubilize hepatocytes can be used as a method to quantify mRNA from hepatocytes. Blood sample lysate stored in guanidine thiocyanate has been shown to maintain nucleic acid integrity. Research describes guanidine thiocyanate to be ineffective for RNA isolation of plant tissue with a high levels of secondary metabolites and storage proteins.

Soluble in H₂O (1420 g/L) at 20°C, and ethanol (5%).

Achtung! Bitte beachten: Light Sensitive. Keep container tightly closed and store away from oxidising agents.


Formel: CH₅N₃·CHNS
Melting Pt: 118…122 °C
Dichte: 1,29
Storage Temperature: Raumtemperatur
MDL Number: MFCD00013027
CAS-Nummer: 593-84-0
EINECS: 209-812-1
Order Now
Spezifikation Testergebnisse
Appearance (Colour) White to off-white
Form Crystalline powder
Assay (Titration) 99.0% min.
DNase None detected
Protease activity None detected
RNase None detected
UV/Vis A280 (70%, water) 0.8
UV/Vis A300 (6M, water) 0.1
UV/Vis A410 (6M, water) 0.1

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge