J65939.MA
J65939.MAEA
114
EUR
InStock
J65939.MA
CPTH2
CPTH2
A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators.
Brown Solid
Applications: A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators
Soluble in DMSO