J66582.06
J66582.06EA
98.8
EUR
InStock
J66582.06
Etionamid 98%
Ethionamide
Purity: 98%
CAS No.: 536-33-4
Molecular Formula: C8H10N2S
Formula Weight: 166.24
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Physical Form: Powder
Appearance: Yellow
Melting Point: 160-162° dec.
Boiling Point: 297°
Solubility: Soluble in DMSO. Sparingly soluble in water
MDL No.: MFCD00057361