J66615.MC
J66615.MCEA
220
EUR
InStock
J66615.MC
Flucloxacillin sodium
Flucloxacillin sodium
Targets primarily the cell wall of susceptible Gram-positive bacteria including Staphylococcus aureus.
CAS No.: 1847-24-1
Molecular Formula: C19H16ClFN3NaO5S
Formula Weight: 475.85
Flash Point: 110°(230°F)
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White
Melting Point: 176-178°
Boiling Point: 677°
Solubility: Soluble in water
MDL No.: MFCD01682108
Applications: Targets primarily the cell wall of susceptible gram positive bacteria including Staphylococcus aureus.