Anhydrotetracycline hydrochloride
Lieferant: ALFA AESAR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
J66688.MA
J66688.MAEA 77.5 EUR InStock
J66688.MA
Anhydrotetracycline hydrochloride
Anhydrotetracycline hydrochloride
Anhydrotetracycline hydrochloride acts as a useful effector of tetracycline-dependent gene expression in bacterial expression systems.
Achtung! Bitte beachten: Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Formel: C₂₂H₂₃ClN₂O₇
Melting Pt: 220…223 °C
Storage Temperature: Gefrierschrank
MDL Number: MFCD00151453
CAS-Nummer: 13803-65-1
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge