Multi-parameter meter (pH/conductivity), bench, 3540
Lieferant: Cole-Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
354001 79000-58
662-0331EA 1750 EUR InStock
662-0331 JENW79000-58
Multi-parameter meter (pH/conductivity), bench, 3540
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
Ideal for use in all laboratories where pH and conductivity analyses are required.
  • Simultaneous display of pH and conductivity
  • Two independent channels to prevent interference between probes
  • Additional modes for resistivity, salinity, TDS and mV
  • Data logger with auto save/print options
  • Storage of up to 500 readings (250 for each mode)
Bestellinformation: Supplied with glass combination pH electrode 662-0343, glass conductivity probe (K=1, 663-0005), electrode stand and holder, ATC probe 662-0338, BNC shorting plug, pH buffers and power supply.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge