Jenway® Cuvettes
Lieferant: Cole-Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
83070-40 83070-34 99610-33 83070-76
JENW83070-40EA 243 EUR InStock
JENW83070-40 JENW83070-34 JENW99610-33 JENW83070-76
Jenway® Cuvettes
Küvetten Spektralphotometer-Küvetten
Get accurate spectroscopic readings.
  • Obtain consistent spectrophotometric data
Quartz and glass cuvettes are constructed of durable materials, making them ideal for repeated use. Single-use polystyrene cuvettes provide fast spectroscopic analysis and offer a less expensive alternative to glass and quartz cuvettes while eliminating washing and potential cross-contamination.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge