Cuvettes for spectrophotometers
Lieferant: Cole-Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
83070-42 99610-31 83070-50 99610-30 99610-32 83070-80 83070-82
JENW83070-42EA 559 EUR InStock
JENW83070-42 JENW99610-31 JENW83070-50 JENW99610-30 JENW99610-32 JENW83070-80 JENW83070-82
Cuvettes for spectrophotometers
Küvetten Spektralphotometer-Küvetten
Extensive range of cuvettes for spectrophotometry for measuring liquids.
Glass cuvettes are designed for the visible region whereas quartz cuvettes may be used in both the UV and visible regions.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge