Safety caps, DIN 50 threads
Lieferant: VICI JOUR RESEARCH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
JR-S-15012 JR-S-15011
JOURJR-S-15012EA 314 EUR InStock
JOURJR-S-15012 JOURJR-S-15011
Safety caps, DIN 50 threads
Verschlüsse Safety Caps und Safety Waste Caps
DIN50 safety caps seal to the bottle/canister with an O-ring behind a PTFE lip, so the O-ring is therefore not a wetted part. Unused ports can be closed with the stopcock.
  • Built-in stopcocks for easy interruption of solvent lines
  • Fit all bottles or canisters with DIN50 thread
  • Prevent evaporation of volatile compounds
  • Stopcocks rated up to 4,96 bar gas at 25 °C
Bestellinformation: A bottom-sealing barbed adapter to connect 1/8" (3,2 mm) ID tubing can be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge