Chromatography tubing, PFA
Lieferant: VICI JOUR RESEARCH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
JR-T-4001-M3 JR-T-4002-M3 JR-T-4003-M3 JR-T-4007-M3 JR-T-4007-M25 JR-T-4007-M10 JR-T-4003-M25 JR-T-4003-M10 JR-T-4002-M10 JR-T-4001-M25 JR-T-4001-M10 JR-T-4002-M25 JR-T-4004
JOURJR-T-4001-M3EA 62.8 EUR InStock
JOURJR-T-4001-M3 JOURJR-T-4002-M3 JOURJR-T-4003-M3 JOURJR-T-4007-M3 JOURJR-T-4007-M25 JOURJR-T-4007-M10 JOURJR-T-4003-M25 JOURJR-T-4003-M10 JOURJR-T-4002-M10 JOURJR-T-4001-M25 JOURJR-T-4001-M10 JOURJR-T-4002-M25 JOURJR-T-4004
Chromatography tubing, PFA
Schläuche Chromatographie-Schläuche
These PFA (perfluoro alkoxy) tubing has excellent chemical stability and mechanical strength.
  • Maximum pressure rating of up to 100 bar
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge