Chromatography tubing, FEP
Lieferant: VICI JOUR RESEARCH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
JR-T-6320-M3 JR-T-6105-M10 JR-T-6105-M25 JR-T-6105-M3 JR-T-6210-M10 JR-T-6210-M25 JR-T-6210-M3 JR-T-6310-M10 JR-T-6310-M25 JR-T-6310-M3 JR-T-6320-M10 JR-T-6605C-M10 JR-T-6620C-M10 JR-T-6620C-M25 JR-T-6620C-M3 JR-T-6620-M1 JR-T-6620-M10 JR-T-6620-M3 JR-T-6615-M10 JR-T-6615-M1 JR-T-6606-M3 JR-T-6605C-M3 JR-T-6605-M1 JR-T-6605-M10 JR-T-6605-M3 JR-T-6606C-M10 JR-T-6606C-M25 JR-T-6606C-M3 JR-T-6615-M3 JR-T-6606-M10 JR-T-6607C-M10 JR-T-6607C-M25 JR-T-6607C-M3 JR-T-6607-M1 JR-T-6607-M10 JR-T-6607-M3 JR-T-6610C-M10 JR-T-6610-M1 JR-T-6610C-M25 JR-T-6610C-M3 JR-T-6610-M10 JR-T-6610-M3 JR-T-6606-M1
JOURJR-T-6320-M3EA 74.5 EUR InStock
JOURJR-T-6320-M3 JOURJR-T-6105-M10 JOURJR-T-6105-M25 JOURJR-T-6105-M3 JOURJR-T-6210-M10 JOURJR-T-6210-M25 JOURJR-T-6210-M3 JOURJR-T-6310-M10 JOURJR-T-6310-M25 JOURJR-T-6310-M3 JOURJR-T-6320-M10 JOURJR-T-6605C-M10 JOURJR-T-6620C-M10 JOURJR-T-6620C-M25 JOURJR-T-6620C-M3 JOURJR-T-6620-M1 JOURJR-T-6620-M10 JOURJR-T-6620-M3 JOURJR-T-6615-M10 JOURJR-T-6615-M1 JOURJR-T-6606-M3 JOURJR-T-6605C-M3 JOURJR-T-6605-M1 JOURJR-T-6605-M10 JOURJR-T-6605-M3 JOURJR-T-6606C-M10 JOURJR-T-6606C-M25 JOURJR-T-6606C-M3 JOURJR-T-6615-M3 JOURJR-T-6606-M10 JOURJR-T-6607C-M10 JOURJR-T-6607C-M25 JOURJR-T-6607C-M3 JOURJR-T-6607-M1 JOURJR-T-6607-M10 JOURJR-T-6607-M3 JOURJR-T-6610C-M10 JOURJR-T-6610-M1 JOURJR-T-6610C-M25 JOURJR-T-6610C-M3 JOURJR-T-6610-M10 JOURJR-T-6610-M3 JOURJR-T-6606-M1
Chromatography tubing, FEP
Schläuche Chromatographie-Schläuche
FEP, natural or coloured.
  • Solid colours identify the tubing Int.Ø.
  • Recommend for ion chromatography applications
FEP (FluorinatedEthylenePropylene) tubing is chemically inert to most HPLC solvents, and is recommended for low pressure applications.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge