Ear defenders, Sonis®
Lieferant: JSP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AEB030-0CY-000 AEB010-0AY-800 AEB020-0AY-900 AEB030-0AY-000 AEB040-0A1-A00
JSPAAEB030-0CY-000EA 53.9 EUR InStock
JSPAAEB030-0CY-000 JSPAAEB010-0AY-800 JSPAAEB020-0AY-900 JSPAAEB030-0AY-000 JSPAAEB040-0A1-A00
Ear defenders, Sonis®
Kapselgehörschützer Lärmschutz-Kapselgehörschützer
The Sonis® ear defender range has been designed with a colour coded system for simple selection, and peaks at an unbeatable 37 SNR.
  • Foam/PET combination cushions
  • ABS cups
  • Optimum cushion pressure and comfort
  • Adjustable steel arms
  • Hygienic
The Sonis® range of ear defenders have been designed by the JSP R&D team. Working with a leading UK acoustic engineering research facility they have developed an ear defender range that peaks at an unbeaten 37 SNR.
The over-moulded headband has a built-in comfort area with ventilation. The headband force and the cushion surface area work together to provide optimum cushion pressure and comfort. The universal slots of JSP helmets allow for the easy fitting of the Sonis® helmet mounted ear defenders. Sonis® cups can be coloured to meet a corporate identity. They can also be printed with a company logo for improved brand recognition and to discourage theft. MOQ's apply.
To facilitate even distribution of pressure the wearer can tilt and adjust the position of the earcup for optimum comfort and fit. The headband is manufactured from TPE, making it easy to wipe clean. The sealing rings are easy to replace. Hygiene kits are available for each model.
Zertifizierungen: EN 352.
Bestellinformation: Available in headband or helmet-mounted versions.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge