Splash guard, without impact protection, Force®8
Lieferant: JSP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AFN060-000-000
JSPAAFN060-000-000EA 27.6 EUR InStock
JSPAAFN060-000-000
Splash guard, without impact protection, Force®8
Gesichtsschutz
The Force®8 Splash Guard can be fitted to a Force®8 half mask to reduce the likelihood of exhaled bacteria/virus hitting the wearer's face.
  • Easily fits to JSP Force®8 half mask
  • Provides a layer of protection to block aerosols and droplets
  • Prevents wearer from touching eyes and face
  • Quick and simple cleaning with soap and water
The Force®8 Splash Guard prevents the wearer from touching eyes and face. Quick and simple cleaning can be performed with soap and water.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge