Dust and liquid goggles, Martcare®
Lieferant: JSP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AGC020-301-300 AGC021-201-300
JSPAAGC020-301-300EA 6 EUR InStock
JSPAAGC020-301-300 JSPAAGC021-201-300
Dust and liquid goggles, Martcare®
Vollsichtbrillen
A cost-effective, compact, and lightweight goggle.
  • Compact and lightweight
  • Tough polycarbonate lens
  • Impact protection
  • Indirect venting offers dust and liquid protection
  • PVC frame
A compact and lightweight goggle with tough polycarbonate lens and PVC frame, offering high resistance to impact from flying particles.
Zertifizierungen: Conforms to EN 166 1.B.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge