Industrial working at height safety helmet, EVOLite® Skyworker™
Lieferant: JSP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AJS260-000-100
JSPAAJS260-000-100EA 129 EUR InStock
JSPAAJS260-000-100
Industrial working at height safety helmet, EVOLite® Skyworker™
Schutzhelme Schutzhelme
The EVOLite® Skyworker™ helmet has a suspension system and shell structure that is designed for use in industrial, mountaineering, rescue, and leisure activities.
  • Industrial working at height safety helmet
  • Meets the EN 12492 mountaineering standard
  • Strong lightweight ABS shell
  • EPS liner for side, front, and rear impact protection
  • Fully adjustable 4-point chinstrap with quick release buckle
The EVOLite® Skyworker™ helmet conforms to the mandatory shock absorption and penetration requirements of EN 397 industrial standard. The helmet features a fully adjustable 4-point chinstrap with quick release buckle. Side, front and rear impact protection are provided by a shock-absorbing EPS liner. The liner is vented to increase comfort whilst maintaining a high protection factor.
The helmet also features a 6-point Terylene harness system and Chamlon™ sweatband. Revolution® wheel ratchet, 3-D adjustment, and a vented shell reduce heat within the helmet.
Zertifizierungen: EN 397, EN 12492.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge