Olympus® safety helmet, EVO®5
Lieferant: JSP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AKF170-000-100
JSPAAKF170-000-100EA 50.7 EUR InStock
JSPAAKF170-000-100
Olympus® safety helmet, EVO®5
Schutzhelme Schutzhelme
The EVO®5 Olympus® safety helmet combines a super strong ABS shell designed for superior all day protection with the comfort benefits of the Evolution® 3D Adjustment™ harness system. The helmet passes all relevant industrial international standards.
  • Global multi-standard hard hat
  • Tough ABS shell
  • Vented design
  • 6-point Terylene cradle harness system
  • Revolution® wheel ratchet adjustment
The EVO®5 Olympus® safety helmet features a 6-point terylene cradle harness system and an Egyptian cotton Chamlon™ sweatband that offers unrivalled comfort. 3-D adjustment offers 1-2-3 point harness depth settings for a perfect fit. The Revolution® wheel ratchet has deep nape strap that enables the firmest retention of any helmet, with a simple turn fastening system.
Side and rear ventilation for optimum air movement inside the shell, reducing helmet temperatures by an average 2 to 3 degrees, increasing the comfort for the wearer. The textile harness features unique 1-2-3 point harness depth settings. This design provides the most precise fit attainable on an industrial safety helmet.
Zertifizierungen: EN 397; ANSI / ISea Z89.1; CSA Z94.1; GB2811; AS/NZS1801.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge