Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Disposable moulded mask, Martcare®

Lieferant: JSP

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BEG120-002-B00 BEG130-002-B00 BEH120-002-A00 BEH130-002-A00
JSPABEG120-002-B00EA 10.6 EUR InStock
JSPABEG120-002-B00 JSPABEG130-002-B00 JSPABEH120-002-A00 JSPABEH130-002-A00
Disposable moulded mask, Martcare®
Atemschutzmasken Einweg-Atemschutzmasken Atemschutzmasken gegen Partikel
A range of polypropylene moulded disposable respirators, available in valved and unvalved options, offering a cost effective solution.

  • Inner foam nose bridge
  • Metal-free straps
  • Exhalation valve option
  • Cost effective

The Martcare® range of polypropylene moulded disposable respirators offers a cost-effective solution, subject to the rigorous batch testing and conformity procedures applicable to the entire JSP PPE range.

Mask straps are ultrasonically welded, ensuring no metal parts come into contact with the face. A foam area around the nose of the mask provides comfort and a firmer fit. A low resistance exhalation valve option is available for easy breathing.

FFP2 options offer 94% minimum filtration efficiency. FFP3 options offer 99% minimum filtration efficiency.

Zertifizierungen: EN 149:2001+A1:2009.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge