Half mask with PressToCheck™ P3 R D filters, Force®8, JSP LTD
Lieferant: JSP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BHT0A3-0L5-N00
JSPABHT0A3-0L5-N00EA 67 EUR InStock
JSPABHT0A3-0L5-N00
Half mask with PressToCheck™ P3 R D filters, Force®8, JSP LTD
Atemschutzmasken Mehrweg-Atemschutzmasken Halbmasken
The Force®8 twin cartridge half mask with Typhoon™ valve offers superior low breathing resistance and a 4-point suspension harness with quick-release buckles, fitted with P3 PressToCheck™ filters for daily face fit reassurance.
  • Durable thermoplastic rubber mask for superior fit to most face shapes
  • Fully adjustable 4-point cradle suspension harness
  • Low resistance exhalation valve
  • Fitted with PressToCheck™ P3 R D filters
  • CR2 reflectivity
The Force®8 twin cartridge half mask with Typhoon™ valve is made with a durable thermoplastic rubber, offering a superior fit to most face shapes. The mask is also fitted with PressToCheck™ P3 R D filters for daily face fit reassurance.
CR2 reflective strips create increased visibility in low-light environments for added safety.
Zertifizierungen: Conforms to EN 140 & EN 143:200.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge