Laboratory jacks
Lieferant: JUCHHEIM Laborgeräte GmbH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VL02280 VL02290 VL02291 VL02291/3 VL02292 VL02293
JUCHVL02280EA 63.42 EUR InStock
JUCHVL02280 JUCHVL02290 JUCHVL02291 JUCHVL02291/3 JUCHVL02292 JUCHVL02293
Laboratory jacks
Laborboy
Aluminium alloy, with epoxy resin coating. These manually operated lifting stages provide high stablity.
  • Special epoxy resin coating ensures that the surfaces are extremely resistant to chemical attack and to other environmental factors
  • Strict quality controls, as required by DIN EN ISO 9001:2000, ensure a constant product quality
  • Made in Germany
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge