Bosshead, square
Lieferant: JUCHHEIM Laborgeräte GmbH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VL04809VA
JUCHVL04809VAEA 10.5 EUR InStock
JUCHVL04809VA
Bosshead, square
Klemmen Stativmuffen
Bosshead made from tempered cast iron, epoxy coated, with stainless steel hex socket (M6).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge