Recirculating coolers, SemiChill series
Lieferant: JULABO GmbH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
950005007P3H0 950005107P3H0 950010107P3H0 950002603P3H0 950002503P3H0
JULA950005007P3H0EA 16850 EUR InStock
JULA950005007P3H0 JULA950005107P3H0 JULA950010107P3H0 JULA950002603P3H0 JULA950002503P3H0
Recirculating coolers, SemiChill series
Umwälzthermostate Geschlossene Umwälzsysteme Kälteumwälzthermostate
Recirculating coolers can handle virtually any cooling requirements in laboratories or industrial environments efficiently.
  • Environmentally friendly operation with low energy consumption
  • Large, bright LED display with splash-proof keypad
  • Optional air or water cooling with easy filling and drain tap easily accessible
  • Circulating pumps with flow rates up to 60 L/min and 6 bar pressure
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge