Safety shelf cans, nonmetallic
Lieferant: JUSTRITE MANUFACTURING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
14169
JUSU14169EA 507 EUR InStock
JUSU14169
Safety shelf cans, nonmetallic
Sicherheitsbehälter
Polyethylene.
  • Nonmetallic shelf cans have a faucet that is mounted two inches above the bottom to eliminate lifting the cans during dispensing
  • Nonmetallic tilt cans have faucet on top and the can rests in a sturdy metal cradle
  • Current-carrying carbon insert imbedded into the container provides proper grounding
  • Stainless steel hardware on the top of can provides maximum corrosion resistance and long life
These nonmetallic safety cans are ideal for dispensing the most corrosive liquids. It has a self-closing faucet for easy dispensing and all fill spouts have a pressure relief cap with stainless steel flame arrester.
Zertifizierungen: NFPA, OSHA and FM approved
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge