Countertop PE Cabinet for Corrosives
Lieferant: JUSTRITE MANUFACTURING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
24080
JUSU24080EA 917 EUR InStock
JUSU24080
Countertop PE Cabinet for Corrosives
Möbel Lagerschränke
Store acids or bases in a completely metal-free countertop polyethylene cabinet - never worry about corrosion again.
  • Store two, 4-liter bottles
  • Completely metal free polyethylene design
  • Hinge door from either side for best worktop utilization
  • Choose from one of three included application specific labels - ACID, BASE, CORROSIVE
  • Includes two polyethylene spill trays - one for inside the cabinet and one for the worktop
Safely store up to two 4-liter bottles of acids or bases in this durable, compact cabinet. Never worry about corrosion - completely metal free polyethylene design offers excellent chemical resistance against spills or damaging vapours. Includes two polyethylene spill trays, use one for inside the cabinet to contain leaks and remove for easy cleaning. Store the extra tray under the cabinet and remove for use as a handy work top. Hinge door from either side for best worktop utilization - accepts a padlock (not included) for security. Three application-specific labels identify contents to safety store and segregate incompatible chemicals - ACID, BASE, and CORROSIVE.
Bestellinformation: Accessories not included.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge