Skywalker® 453 II PUR Anti-static Mat ESD
Lieferant: JUSTRITE MANUFACTURING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
453-KNIEMATTE
JUSU453-KNIEMATTEEA 38.8 EUR InStock
JUSU453-KNIEMATTE
Skywalker® 453 II PUR Anti-static Mat ESD
Fußmatten Permanente Bodenmatten
High-quality polyurethane foam anti-fatigue mats are reputed for their ultimate comfort in stationary standing positions. Its key characteristic of high thermal insulation performance is owed to a uniform closed cell structure wherein gas remains trapped.
  • Medium duty
  • R11 anti-slip pebble pattern
  • ESD static dissipative
  • Available in six standard sizes
  • Made from polyurethane foam
The integral skin adds to the compression force deflection properties (>120 according to ASTM 3574C), provides resistance to wear and is non-porous. Ideal for stationary standing workstations, sit-stand desks, packing stations, office environments, reception areas, wait stations, service counters, cashiers.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge