Pipette controller, Seropet
Lieferant: KARTELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1295.10
KART1295.10EA 1880 EUR InStock
KART1295.10
Pipette controller, Seropet
Pipetten Pipettierhelfer
Seropet was developed for aspirating liquids by glass or plastic pipettes ranging from the volume of 0,5 ml up to 100 ml.
  • Two different work modes: quick and normal, with continuous or pulsating adjustment
  • Particularly light structure ensures comfortable use even when used for a long time
  • Supplied with safety valve to block aspiration excesses
  • Equipped with autoclavable protective hydrophobic anti-aerosol filter
  • Highly sensitive internal valves to get precision also with small volumes
  • NiMH battery supplied has an autonomy of up to 8 hours with continuous work and up to 1000 charge cycles
  • Pipette housing is autoclavable
Lieferumfang: Supplied with battery charger, protective PTFE hydrophobic filter (0,2 µm) and an adhesive support for attachment to the laboratory bench.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge