Ballonflaschen
Lieferant: KARTELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1781 1782 1783 1780
KART1781EA 635 EUR InStock
KART1781 KART1782 216-1040 KART1780
Ballonflaschen
Flaschen Kanister
HDPE, translucent with black screw cap.
  • With comfortable handle and secure seal cap following DIN recommendations
  • Conform to UN regulation on transportation of dangerous liquids
  • Suitable for foodstuff
Ideal for storing and transporting liquids.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge