Lever pumps for viscous liquids, MR60
Lieferant: KARTELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2004
KART2004EA 480 EUR InStock
KART2004
Lever pumps for viscous liquids, MR60
Pumpen Fass/Transferpumpen
Dispensing pump is operated by a unique lever-action stroke and can handle the most viscous materials.
  • Spigot orientation of 360° allows versatility in dispensing
  • Complete with shank
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge