Metal trays for histology
Lieferant: KARTELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2923 2924
KART2923EA 117 EUR InStock
KART2923 KART2924
Metal trays for histology
Formen Histologie-Formen
Made of stainless steel for easy removal of paraffin inclusions: they allow a precise location of the specimen into the paraffin.
Verpackung: Packed in bags of 10 pieces.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge