Etiketten
Lieferant: KARTELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5011 5010 5015 5012 5013 5014 5016 5018
KART5011EA 415 EUR InStock
KART5011 KART5010 KART5015 KART5012 KART5013 KART5014 KART5016 KART5018
Etiketten
Etiketten
Transparent, punched labels for use with wash bottles and plastic or glass reagent bottles.
  • Complete with risk assessment numbers
  • Recommended by the NFPA (National Fire Protection Association)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge