Dishes
Lieferant: KARTELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
625 627
KART625EA 63.5 EUR InStock
KART625 KART627
Dishes
Schalen/Schüsseln/Töpfe Abdampfschalen
Made of PTFE, white.
  • Very good chemical resistance
  • Very good temperature resistance
  • Good mechanical resistance
Dishes are suitable for foodstuff. It can withstand temperatures continuously up to 250 °C and for short intervals up to 300 °C.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge