Pistille, geriffelt, PTFE-Kopf
Lieferant: KARTELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6315 6302 6305 6310 6330 6350
KART6315EA 115 EUR InStock
KART6315 KART6302 KART6305 KART6310 KART6330 KART6350
Pistille, geriffelt, PTFE-Kopf
Mörser und Pistille Pistille
Pestles used in the combination with borosilicate mortar for sample crushing. Ideal for tough materials such as tumours.
  • Head made of PTFE
  • Shaft made of stainless steel
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge