Wash bottles, narrow neck
Lieferant: Kautex
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
76082 76084
KAUT76082EA 2.5 EUR InStock
KAUT76082 KAUT76084
Wash bottles, narrow neck
Flaschen Spritzflaschen
LDPE, translucent with LDPE spray cap.
  • Good chemical resistance
  • Heat resistant up to 80 °C
  • Leakproof
Economical, soft and very easy to press. For use with general solutions such as water, soap, dilute acids and dilute alkalis. The tip can be cut off to increase the flow rate.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge