Dishes, evaporating, flat or round Bottom, with spout
Lieferant: Kavalier
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
632411629060 632411604050 632411604100 632411604130 632411604160 632411604200 632411604230 632411629115 632411629070_U 632411629095 632411629140 632411604060 632411629070
216-1192EA 27.5 EUR InStock
216-1192 KAVA632411604050 KAVA632411604100 KAVA632411604130 KAVA632411604160 KAVA632411604200 KAVA632411604230 KAVA632411629115 KAVA632411629070_U KAVA632411629095 KAVA632411629140 KAVA632411604060 KAVA632411629070
Dishes, evaporating, flat or round Bottom, with spout
Schalen/Schüsseln/Töpfe Abdampfschalen
Made of borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For a wide range of applications. With flat bottom or rund bottom.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge