Automatische Pipetten, Kipp, mit Flasche
Lieferant: Kavalier
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
632446003005
KAVA632446003005EA 3490 EUR InStock
KAVA632446003005
Automatische Pipetten, Kipp, mit Flasche
Kippautomaten
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
This dispenser head with standard ground jont NS 29/32 is completed by a 250 ml flask for repetitive dispensing of the same volume.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge