General purpose gloves, Ideal 752
Lieferant: KCL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
752.8
KCHL752.8EA 361 EUR InStock
KCHL752.8
General purpose gloves, Ideal 752
Handschuhe
These natural latex gloves with flock lining provide protection for hand and materials during routine work in canteens, cleaning and hygiene environments.
  • Excellent grip
  • Easier donning
  • Good dexterity
  • Prevents user fatigue
  • High wearing comfort thanks to flock lining
  • Special follow-up treatment process for a low emission of natural latex particles
Zertifizierungen: Cat. I
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge