Hanging balances, HCB and HCN Series
Lieferant: KERN SOHN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
HCB20K10
KERNHCB20K10EA 120 EUR InStock
KERNHCB20K10
Hanging balances, HCB and HCN Series
Waagen
Ideal for rapid checking in goods receiving and shipping.
  • Data hold function: For easy reading of the weight, the display can be 'frozen' in different ways, either automatically when the weighing value remains unchanged or manually by pressing the Hold key
  • Peak load display (peak hold), measuring frequency 5 Hz
Zertifizierungen: Please note: If a calibration certificate is required, this should be requested when ordering the balance, as it can not be issued retrospectively.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge