Polytron® PTG 36/4 Generators
Lieferant: KINEMATICA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9810065
KINE9810065EA 0 EUR InStock
KINE9810065
Polytron® PTG 36/4 Generators
Homogenisatoren Dispergierwerkzeuge
Standard generator rotor/stator with two teeth rows each.
  • Making fine emulsions and suspensions
  • Mixes and reduces highly intensive solids into fluids
  • Used for gaseous solutions and extraktions
  • Separates fibres and cellular material into very small particles
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge