Cold trap with scealing cap
Lieferant: KNF
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
057955
KNFA057955EA 0 EUR InStock
KNFA057955
Cold trap with scealing cap
Vakuumfallen
Cold trap made of borosilicate glass 3.3, with GL 45 closing cap.
  • Features hose with threaded caps GL 18
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge