Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Vacuum control unit, VC 900

Lieferant: KNF

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

VC900
KNFAVC900EA 2573 EUR InStock
KNFAVC900
Vacuum control unit, VC 900
Pumpen Vakuum-Controller
Vacuum control unit to control the vacuum applications, like rotary evaporation, distillation, multi-user vacuum system.

  • Easy to use to control the vacuum application
  • Separate control unit with pressure sensors and two-step controlled valve to be placed independently from the operating unit
  • Vacuum controller can be operated with components from KNF and other producers

When used in combination with KNF diaphragm vacuum pump N 920 G, the two-step control of the VC 900 switches to a precise rotational speed control by means of the pump’s vacuum control unit interface.

Housing: plastic

Specifications of the Control Unit:
Hose connections (mm) for gas in: ID 10, PVDF
Hose connections (mm) for gas out: ID 10, PVDF
Hose connections (mm) for venting ID 4, nickel-plated brass
Dimensions WxDxH (mm): 155×60×109 mm

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge