Digital Indicator for Field Housing, 830XR
Lieferant: KNICK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
830XR
KNIC830XREA 0 EUR InStock
KNIC830XR
Digital Indicator for Field Housing, 830XR
Elektrochemische Messgeräte Leitfähigkeitsmessgeräte
Digital loop-powered 830 series indicators are universally useable in current measurement circuits, switching (either 0 to 20 mA or 4 to 20 mA) is simply performed via the terminals.
  • Digital indicator without power supply or power cables
  • Indicator not affected by grid outage
  • Universal range adjustment
  • Selectable unit symbols
  • Potential isolation Ex/Ex
  • Voltage drop 0,5 V
  • Large 23-mm digital numbers
  • 3-year warranty
The measured value is displayed for a current as low as 0,3 mA. Thanks to the low voltage drop of just 0,5 V, the use in current loops with a low load voltage is also possible.
4-Digit display: measuring range up to 10000, display range −9999 to +9999
Range overview through additional integrated bargraph, Configurable without external reference current, Parameters can also be modified during operation, Settings protected by a passcode, Approved for use in HART circuits, No signal interference from power supply coupling and No parasitic voltages.
Zertifizierungen: IP 65
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge