Racks for microtubes and cryo vials
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2-1642
KOEH2-1642EA 27.1 EUR InStock
KOEH2-1642
Racks for microtubes and cryo vials
Gestelle Gestelle für Kryoröhrchen
PP rack, square, easy to mount. For 1,5/2,0 ml mircrocentrifuge tubes and cryogenic vials.
  • Alphanumeric index of the holes
  • Solid bottom of each well to secure vials in place for easier opening and closing
  • Fits perfectly and is stackable even when filled with tubes
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge