Beta Radiation Waste Boxes
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A-1123
KOEHA-1123EA 679 EUR InStock
KOEHA-1123
Beta Radiation Waste Boxes
Boxen Beta-/Gammastrahlen-Sicherheitsbehälter
Beta radiation waste disposal boxes, acrylic, with tightly closing hinged lid.
  • Suitable for disposal of other critical materials
  • Lid provides protection against contaminated materials
Material thickness: 10 mm
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge