Nitrate/ammonium standard solution
Lieferant: KRAFT BERND
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
27278.2000
KRAF27278.2000EA 102 EUR InStock
KRAF27278.2000
Nitrate/ammonium standard solution
Standards Anionische Standards Nitrat-Standards
Solution with 100 mg each NH₄-N + NO₃-N/l KNO₃ + (NH₄)₂SO₄ in water for TNb determination.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge