Manual polarimeter
Lieferant: A.KRÜSS Optronic
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
P1000-LED
KRUEP1000-LEDEA 1820 EUR InStock
KRUEP1000-LED
Manual polarimeter
Polarimeter
This manual polarimeter features a high-quality metal stand and a sample chamber.
  • Very low power consumption
  • Reading is done via the eyepiece with two verniers
Suitable for all applications in the laboratory on the principle of partial shade.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge