Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Trousers for women, C.I., Design c, black

Lieferant: FRISTADS KANSAS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

844940131C44 844940131C46 844940131C48 844940210C44 844940210C46 844940210C48 844940231C44 844940231C46 844940231C48 844940331C44 844940331C46 844940331C48 844940440C44 844940440C46 844940440C48 844940520C44 844940520C46 844940520C48 844940530C44 844940530C46 844940530C48 844940539C44 844940539C46 844940539C48 844940640C44 844940640C46 844940640C48 844940730C44 844940730C46 844940730C48 844940940C44 844940940C46 844940940C48 844940941C44 844940941C46 844940941C48 84494034 84494036 84494038 84494040 84494042 84494044 84494046 84494048 84494050
KWIN844940131C44EA 0 EUR InStock
KWIN844940131C44 KWIN844940131C46 KWIN844940131C48 KWIN844940210C44 KWIN844940210C46 KWIN844940210C48 KWIN844940231C44 KWIN844940231C46 KWIN844940231C48 KWIN844940331C44 KWIN844940331C46 KWIN844940331C48 KWIN844940440C44 KWIN844940440C46 KWIN844940440C48 KWIN844940520C44 KWIN844940520C46 KWIN844940520C48 KWIN844940530C44 KWIN844940530C46 KWIN844940530C48 KWIN844940539C44 KWIN844940539C46 KWIN844940539C48 KWIN844940640C44 KWIN844940640C46 KWIN844940640C48 KWIN844940730C44 KWIN844940730C46 KWIN844940730C48 KWIN844940940C44 KWIN844940940C46 KWIN844940940C48 KWIN844940941C44 KWIN844940941C46 KWIN844940941C48 KWIN84494034 KWIN84494036 KWIN84494038 KWIN84494040 KWIN84494042 KWIN84494044 KWIN84494046 KWIN84494048 KWIN84494050
Trousers for women, C.I., Design c, black
Arbeitskleidung
Robust trousers for ladies, made from 65% polyester and 35% cotton, with soft nap on reverse side.

  • Fly with zip
  • Two front pockets, two back pockets with flap and velcro closure
  • Leg pockets with flaps and three tool pockets, leg pocket with mobile phone pocket and two pen pockets
  • Folding-ruler pocket, D-ring, inside knee pockets

Weight: 260 g/m²

Basic colour: Black

Bestellinformation: Trousers are available without any coloured accents or with 12 different accent colours.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge