L00355.09
L00355.18
L00355.09EA
94.8
EUR
InStock
L00355.09
L00355.18
N-Ethylmaleimid (NEM) ≥98%
N-Ethylmaleimid (NEM)
Purity: 98+%
CAS No.: 128-53-0
Molecular Formula: C6H7NO2
Formula Weight: 125.13
Flash Point: 73°(163°F)
Odour: Pungent
Boiling Point: 209-211°
Beilstein Reference: 112448
Merck Reference: 14,3822
UN No.: 2928
MDL No.: MFCD00005509