Cell culture media, Basal Medium Eagle (BME)
Lieferant: Biowest
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
L0042-500
L0042-500EA 14.4 EUR InStock
L0042-500
Cell culture media, Basal Medium Eagle (BME)
Zellkulturmedien
Basal Medium Eagle is principally used for monolayer diploid or primary cell cultures. It is the simplest of the basal media with all essential components for growth. BME ideally supports cell lines such as HeLa, L-cells and primary mammalian fibroblasts. BME has also successfully been used for the growth of human transformed cell lines (particularly WI-38 and MRC-5).
Achtung! Bitte beachten: Please note that BME contains Earle's salts as a buffer, it should therefore be used in an environment containing 5 to 10% CO₂ in order to maintain physiological pH.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge