L00582.06
L00582.14
L00582.22
L00582.06EA
25.6
EUR
InStock
L00582.06
L00582.14
L00582.22
Ethylbrompyruvat ≥75%, technische Qualität
Ethylbrompyruvat
Ethyl 3-bromopyruvate is employed in a synthesis of thioxothiazolidines from primary amines and carbon disulfide
Purity: 85%
CAS No.: 70-23-5
Molecular Formula: C5H7BrO3
Formula Weight: 195.02
Flash Point: 98°(208°F)
Refractive Index: 1.4690
Boiling Point: 98-100°/10mm
Beilstein Reference: 1760158
UN No.: 3265
MDL No.: MFCD00000204